accessibility menu, dialog, popup

UserName

H3CNNH2NNH2N

CAS RN 2533-34-8

IUPAC Name: 6-undecyl-1,3,5-triazine-2,4-diamine
Synonyms: 6-undecyl-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol): 265.41
Molecular Formula: C14H27N5
InChi Key: MLCIKWISJBFZKS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC1=NC(N)=NC(N)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results