accessibility menu, dialog, popup

UserName

CAS RN 2524-49-4

CH2H2N

CAS RN 2524-49-4

IUPAC Name: but-3-en-1-amine
Synonyms: 3-buten-1-amine
Molecular Weight (g/mol): 71.12
Molecular Formula: C4H9N
InChi Key: ASVKKRLMJCWVQF-UHFFFAOYSA-N
SMILES: NCCC=C
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)