accessibility menu, dialog, popup

UserName

HOOHN

CAS RN 2517-04-6

IUPAC Name: azetidine-2-carboxylic acid
Synonyms: azetidine-2-carboxylic acid
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: IADUEWIQBXOCDZ-UHFFFAOYNA-N
SMILES: OC(=O)C1CCN1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)