accessibility menu, dialog, popup

UserName

NHOOH

CAS RN 25137-00-2

IUPAC Name: piperidine-3-carboxylic acid
Synonyms: nipecotic acid
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: XJLSEXAGTJCILF-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCNC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)