accessibility menu, dialog, popup

UserName

OHOClBr

CAS RN 25118-59-6

IUPAC Name: 4-bromo-3-chlorobenzoic acid
Synonyms: 4-bromo-3-chlorobenzoic acid
Molecular Weight (g/mol): 235.46
Molecular Formula: C7H4BrClO2
InChi Key: PSKJIHDVFDVNBU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results