CAS RN 25103-58-6
IUPAC Name:
2,3,3,4,4,5-hexamethylhexane-2-thiol
Synonyms:
2,3,3,4,4,5-hexamethylhexane-2-thiol
Molecular Weight (g/mol):
202.4
Molecular Formula:
C12H26S
InChi Key:
YAJYJWXEWKRTPO-UHFFFAOYSA-N
SMILES:
CC(C)C(C)(C)C(C)(C)C(C)(C)S
1
–
4
of
4
results
1
tert-Dodecanethiol (Mixture of Isomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
tert-Dodecanethiol (Mixture of Isomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
tert-Dodecanethiol (Mixture of Isomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.