accessibility menu, dialog, popup

UserName

CAS RN 2510-32-9

IUPAC Name: 4-methyl-1,3-oxazole-5-carboxylic acid
Synonyms: 4-methyl-1,3-oxazole-5-carboxylic acid
Molecular Weight (g/mol): 127.1
Molecular Formula: C5H5NO3
InChi Key: ZIXUNDOOBLSXPE-UHFFFAOYSA-N
SMILES: CC1=C(OC=N1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results