CAS RN 25067-59-8
CAS RN 25067-59-8
IUPAC Name:
9-ethenyl-9H-carbazole
Synonyms:
N-vinylcarbazole
Molecular Weight (g/mol):
193.25
Molecular Formula:
C14H11N
InChi Key:
KKFHAJHLJHVUDM-UHFFFAOYSA-N
SMILES:
C=CN1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
2
of
2
results
1
Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400
CAS: 25067-59-8 Molecular Formula: (C14H11N)n Molecular Weight (g/mol): 193.25 MDL Number: MFCD00134336 InChI Key: KKFHAJHLJHVUDM-UHFFFAOYSA-N Synonym: 9-vinylcarbazole,n-vinylcarbazole,9-vinyl-9h-carbazole,9h-carbazole, 9-ethenyl,n-vinyl carbazole,vinylcarbazole,1-vinylcarbazole,poly 9-vinylcarbazole,carbazole, 9-vinyl,poly-n-vinylcarbazole PubChem CID: 15143 SMILES: *-CC(-*)N1C2=C(C=CC=C2)C2=C1C=CC=C2