accessibility menu, dialog, popup

UserName

CAS RN 2486-66-0

CH3ONOOOOH

CAS RN 2486-66-0

IUPAC Name: 2-ethoxy-4-nitrobenzoic acid
Synonyms: 2-ethoxy-4-nitrobenzoic acid
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H9NO5
InChi Key: BSIOETYGDXDWBR-UHFFFAOYSA-N
SMILES: CCOC1=C(C=CC(=C1)[N+]([O-])=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results