accessibility menu, dialog, popup

UserName

CAS RN 24670-07-3

OOOHHOOOOHHOOOOHHOH2OH2OH2OH2OH2OH2OH2OH2OH2OH2ODyDy

CAS RN 24670-07-3

IUPAC Name: tris(oxalic acid) didysprosium decahydrate
Synonyms: tris(oxalic acid) didysprosium decahydrate
Molecular Weight (g/mol): 775.25
Molecular Formula: C6H26Dy2O22
InChi Key: WJYBVSNZIQHDSA-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O.O.O.O.[Dy].[Dy].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results