accessibility menu, dialog, popup

UserName

NaCH3SiH3CCH3OO2H2H2H2H

CAS RN 24493-21-8

IUPAC Name: sodium 3-(trimethylsilyl)(²H₄)propanoate
Synonyms: sodium 3-(trimethylsilyl)(²H₄)propanoate
Molecular Weight (g/mol): 172.27
Molecular Formula: C6H13NaO2Si
InChi Key: OIIWPAYIXDCDNL-HGFPCDIYSA-M
SMILES: [Na+].[2H]C([2H])(C([O-])=O)C([2H])([2H])[Si](C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)