CAS RN 24404-83-9
CAS RN 24404-83-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glycolithocholic Acid Sodium Salt, TRC
CAS: 24404-83-9 Molecular Formula: C26H42NNaO4 Molecular Weight (g/mol): 455.61 Synonym: Sodium Glycolithocholate,N-(3alpha-Hydroxy-5beta-cholan-24-oyl)glycine Sodium Salt,N-[(3alpha,5beta)-3-Hydroxy-24-oxocholan-24-yl]glycine Sodium Salt; IUPAC Name: sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate SMILES: [Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C