accessibility menu, dialog, popup

UserName

HNN

CAS RN 244-63-3

IUPAC Name: 9H-pyrido[3,4-b]indole
Synonyms: β-carboline norharman norharmane carbazoline
Molecular Weight (g/mol): 168.2
Molecular Formula: C11H8N2
InChi Key: AIFRHYZBTHREPW-UHFFFAOYSA-N
SMILES: N1C2=C(C=CC=C2)C2=C1C=NC=C2
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results