accessibility menu, dialog, popup

UserName

CAS RN 2436-73-9

H3COOOClH3C

CAS RN 2436-73-9

IUPAC Name: methyl 2-(4-chloro-2-methylphenoxy)acetate
Synonyms: methyl 2-(4-chloro-2-methylphenoxy)acetate
Molecular Weight (g/mol): 214.65
Molecular Formula: C10H11ClO3
InChi Key: VWERIRLJUWTNDA-UHFFFAOYSA-N
SMILES: COC(=O)COC1=CC=C(Cl)C=C1C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results