accessibility menu, dialog, popup

UserName

BH3CNOONOOHOOH

CAS RN 24341-76-2

IUPAC Name: (2-methyl-3,5-dinitrophenyl)boronic acid
Synonyms: 2-methyl-3,5-dinitrophenylboronic acid
Molecular Weight (g/mol): 225.95
Molecular Formula: C7H7BN2O6
InChi Key: ZESXGHPFWNSQQV-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=C1B(O)O)[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results