accessibility menu, dialog, popup

UserName

CAS RN 2434-56-2

NH2NH2NN

CAS RN 2434-56-2

IUPAC Name: pyrimidine-4,6-diamine
Synonyms: pyrimidine-4,6-diamine
Molecular Weight (g/mol): 110.12
Molecular Formula: C4H6N4
InChi Key: MISVBCMQSJUHMH-UHFFFAOYSA-N
SMILES: NC1=CC(N)=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results