accessibility menu, dialog, popup

UserName

CAS RN 24324-17-2

IUPAC Name: (9H-fluoren-9-yl)methanol
Synonyms: 9H-fluorene-9-methanol
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: XXSCONYSQQLHTH-UHFFFAOYSA-N
SMILES: OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (7)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)