accessibility menu, dialog, popup

UserName

NHH2NHCl

CAS RN 24214-73-1

IUPAC Name: cyclohexylhydrazine hydrochloride
Synonyms: cyclohexylhydrazine hydrochloride
Molecular Weight (g/mol): 150.65
Molecular Formula: C6H15ClN2
InChi Key: JZRHODNPRNTXKO-UHFFFAOYSA-N
SMILES: Cl.NNC1CCCCC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results