accessibility menu, dialog, popup

UserName

OOOOOOO

CAS RN 2421-28-5

IUPAC Name: 5-(1,3-dioxo-1,3-dihydro-2-benzofuran-5-carbonyl)-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 322.23
Molecular Formula: C17H6O7
InChi Key: VQVIHDPBMFABCQ-UHFFFAOYSA-N
SMILES: O=C(C1=CC=C2C(=O)OC(=O)C2=C1)C1=CC=C2C(=O)OC(=O)C2=C1
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (6)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)