accessibility menu, dialog, popup

UserName

CAS RN 241816-11-5

NNO

CAS RN 241816-11-5

IUPAC Name: 6-(piperidin-1-yl)pyridine-3-carbaldehyde
Synonyms: 6-(piperidin-1-yl)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 190.25
Molecular Formula: C11H14N2O
InChi Key: RKIMIISNVCLRLA-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(C=C1)N1CCCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)