accessibility menu, dialog, popup

UserName

CAS RN 24044-07-3

IUPAC Name: 2-(thiophen-2-yl)-1,3-thiazole-4-carboxylate
Synonyms: 2-(thiophen-2-yl)-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 210.25
Molecular Formula: C8H4NO2S2
InChi Key: FGKCNTGJZXHKFJ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CSC(=N1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results