accessibility menu, dialog, popup

UserName

CAS RN 239097-78-0

IUPAC Name: 5-isocyanato-1-benzothiophene
Synonyms: 5-isocyanato-1-benzothiophene
Molecular Weight (g/mol): 175.21
Molecular Formula: C9H5NOS
InChi Key: GWLZJAWTWVSHCU-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=C2SC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results