accessibility menu, dialog, popup

UserName

FFFSBr

CAS RN 238403-52-6

IUPAC Name: 1-(bromomethyl)-2-[(trifluoromethyl)sulfanyl]benzene
Synonyms: 1-(bromomethyl)-2-[(trifluoromethyl)sulfanyl]benzene
Molecular Weight (g/mol): 271.1
Molecular Formula: C8H6BrF3S
InChi Key: ZSBCQBRVNBLWAW-UHFFFAOYSA-N
SMILES: FC(F)(F)SC1=CC=CC=C1CBr
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results