accessibility menu, dialog, popup

UserName

NHNNNFFF

CAS RN 238403-48-0

IUPAC Name: 1-[4-(trifluoromethyl)pyrimidin-2-yl]-1,4-diazepane
Synonyms: 1-[4-(trifluoromethyl)pyrimidin-2-yl]-1,4-diazepane
Molecular Weight (g/mol): 246.24
Molecular Formula: C10H13F3N4
InChi Key: XVAZHAVSZFQOOQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC(=NC=C1)N1CCCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results