accessibility menu, dialog, popup

UserName

CAS RN 23794-15-2

H3CONCl

CAS RN 23794-15-2

IUPAC Name: 1-(2-chloropyridin-4-yl)ethan-1-one
Synonyms: 1-(2-chloropyridin-4-yl)ethanone
Molecular Weight (g/mol): 155.58
Molecular Formula: C7H6ClNO
InChi Key: ZJCGPQZERFBGSM-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Cl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results