accessibility menu, dialog, popup

UserName

H3CONNH3C

CAS RN 23787-80-6

IUPAC Name: 1-(3-methylpyrazin-2-yl)ethan-1-one
Synonyms: 1-(3-methylpyrazin-2-yl)ethanone
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: QUNOTZOHYZZWKQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=NC=CN=C1C
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (2)
  • (8)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)