accessibility menu, dialog, popup

UserName

FFFClN

CAS RN 23779-97-7

IUPAC Name: 4-chloro-8-(trifluoromethyl)quinoline
Synonyms: 4-chloro-8-(trifluoromethyl)quinoline
Molecular Weight (g/mol): 231.6
Molecular Formula: C10H5ClF3N
InChi Key: LINGICLAECZKAW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C2N=CC=C(Cl)C2=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)