accessibility menu, dialog, popup

UserName

CAS RN 23593-74-0

IUPAC Name: 1-(triphenylmethyl)-1H-1,3-benzodiazole
Synonyms: 1-(triphenylmethyl)-1,3-benzodiazole
Molecular Weight (g/mol): 360.46
Molecular Formula: C26H20N2
InChi Key: KCENPJPWSWRZLS-UHFFFAOYSA-N
SMILES: C1=NC2=CC=CC=C2N1C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results