accessibility menu, dialog, popup

UserName

CAS RN 23585-00-4

CH3NNONHH2N

CAS RN 23585-00-4

IUPAC Name: 1-methyl-1H-imidazole-5-carbohydrazide
Synonyms: 3-methylimidazole-4-carbohydrazide
Molecular Weight (g/mol): 140.15
Molecular Formula: C5H8N4O
InChi Key: CFIAOJVURKEWPG-UHFFFAOYSA-N
SMILES: CN1C=NC=C1C(=O)NN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results