CAS RN 2356-53-8
CAS RN 2356-53-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2-Dichlorotrifluoroethyl Trifluoromethyl Ether, TRC
CAS: 2356-53-8 Molecular Formula: C3OF6Cl2 Molecular Weight (g/mol): 236.93 Synonym: 1,2-Dichloro-1,1,2-trifluoro-2-(trifluoromethoxy)ethane IUPAC Name: 1,2-dichloro-1,1,2-trifluoro-2-(trifluoromethoxy)ethane SMILES: FC(F)(F)OC(F)(Cl)C(F)(F)Cl