accessibility menu, dialog, popup

UserName

CAS RN 2356-16-3

IUPAC Name: ethyl 2-(diethoxyphosphoryl)-2-fluoroacetate
Synonyms: ethyl 2-(diethoxyphosphoryl)-2-fluoroacetate
Molecular Weight (g/mol): 242.18
Molecular Formula: C8H16FO5P
InChi Key: FVPISMANESAJQZ-UHFFFAOYNA-N
SMILES: CCOC(=O)C(F)P(=O)(OCC)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results