accessibility menu, dialog, popup

UserName

CAS RN 23501-93-1

CH3NH3COH

CAS RN 23501-93-1

IUPAC Name: [3-(dimethylamino)phenyl]methanol
Synonyms: [3-(dimethylamino)phenyl]methanol
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: YTUXZVSDDHCTBZ-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC(CO)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results