accessibility menu, dialog, popup

UserName

CH3ClI

CAS RN 23399-70-4

IUPAC Name: 4-chloro-1-iodo-2-methylbenzene
Synonyms: 4-chloro-1-iodo-2-methylbenzene
Molecular Weight (g/mol): 252.48
Molecular Formula: C7H6ClI
InChi Key: ZZNUQVQAJVZRKW-UHFFFAOYSA-N
SMILES: CC1=C(I)C=CC(Cl)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results