CAS RN 23389-75-5
CAS RN 23389-75-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(1-Hydroxyethyl)pyridine, 97%
CAS: 23389-75-5 Molecular Formula: C7H9NO Molecular Weight (g/mol): 123.155 MDL Number: MFCD00077867 InChI Key: HVOAMIOKNARIMR-UHFFFAOYSA-N Synonym: 1-pyridin-4-yl ethanol,4-1-hydroxyethyl pyridine,1-4-pyridyl ethanol,1-pyridin-4-yl-ethanol,1-pyridin-4-yl ethan-1-ol,1-4-pyridinyl ethanol,+/--1-4-pyridyl ethanol,r-4-1-hydroxyethyl pyridine,alpha-methylpyridine-4-methanol,4-pyridylethanol PubChem CID: 90919 IUPAC Name: 1-pyridin-4-ylethanol SMILES: CC(C1=CC=NC=C1)O