accessibility menu, dialog, popup

UserName

HOOHNN

CAS RN 23249-97-0

IUPAC Name: 3-(1H-1,3-benzodiazol-2-yl)propanoic acid
Synonyms: 3-(1H-1,3-benzodiazol-2-yl)propanoic acid
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: XYWJNTOURDMTPI-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=NC2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results