accessibility menu, dialog, popup

UserName

HCOOHNH2

CAS RN 23235-03-2

IUPAC Name: 2-aminopent-4-ynoic acid
Synonyms: propargylglycine
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: DGYHPLMPMRKMPD-UHFFFAOYNA-N
SMILES: NC(CC#C)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)