accessibility menu, dialog, popup

UserName

CAS RN 2318-25-4

HClH3CONHOOCH3

CAS RN 2318-25-4

IUPAC Name: hydrogen ethyl 3-ethoxy-3-iminopropanoate chloride
Synonyms: hydrogen ethyl 3-ethoxy-3-iminopropanoate chloride
Molecular Weight (g/mol): 195.64
Molecular Formula: C7H14ClNO3
InChi Key: HYMXUYQKXCHWDC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCOC(=N)CC(=O)OCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)