accessibility menu, dialog, popup

UserName

H3CNH3COCl

CAS RN 2315-36-8

IUPAC Name: 2-chloro-N,N-diethylacetamide
Synonyms: CDEA
Molecular Weight (g/mol): 149.62
Molecular Formula: C6H12ClNO
InChi Key: CQQUWTMMFMJEFE-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)CCl
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)