accessibility menu, dialog, popup

UserName

CAS RN 2314-36-5

OHClOCl

CAS RN 2314-36-5

IUPAC Name: 3,5-dichloro-4-hydroxybenzaldehyde
Synonyms: 3,5-dichloro-4-hydroxybenzaldehyde
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: LIYGCLJYTHRBQV-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(C=O)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results