accessibility menu, dialog, popup

UserName

CAS RN 2309-48-0

OSS(E)

CAS RN 2309-48-0

IUPAC Name: (2E)-1,3-bis(thiophen-2-yl)prop-2-en-1-one
Synonyms: (2E)-1,3-bis(thiophen-2-yl)prop-2-en-1-one
Molecular Weight (g/mol): 220.3
Molecular Formula: C11H8OS2
InChi Key: WCAGHDMFZMUUPQ-AATRIKPKSA-N
SMILES: O=C(\C=C\C1=CC=CS1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results