accessibility menu, dialog, popup

UserName

CAS RN 2305-25-1

IUPAC Name: ethyl 3-hydroxyhexanoate
Synonyms: ethyl 3-hydroxyhexanoate
Molecular Weight (g/mol): 160.21
Molecular Formula: C8H16O3
InChi Key: LYRIITRHDCNUHV-UHFFFAOYNA-N
SMILES: CCCC(O)CC(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results