accessibility menu, dialog, popup

UserName

CAS RN 23047-25-8

CH3NNOCl

CAS RN 23047-25-8

IUPAC Name: 2-[(3-{2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)(methyl)amino]-1-(4-chlorophenyl)ethan-1-one
Synonyms: lofepramine feprapax lofepramine (substance) 洛非帕明 deftan lopramine gamanil lofepramine (product) rosemont brand of lofepramine hydrochloride merck brand of lofepramine hydrochloride
Molecular Weight (g/mol): 418.97
Molecular Formula: C26H27ClN2O
InChi Key: SAPNXPWPAUFAJU-UHFFFAOYSA-N
SMILES: CN(CCCN1C2=CC=CC=C2CCC2=CC=CC=C12)CC(=O)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results