accessibility menu, dialog, popup

UserName

CAS RN 22978-61-6

ClClNHNH2HCl

CAS RN 22978-61-6

IUPAC Name: 3,5-dichlorobenzene-1-carboximidamide hydrochloride
Synonyms: 3,5-dichlorobenzenecarboximidamide hydrochloride
Molecular Weight (g/mol): 225.5
Molecular Formula: C7H7Cl3N2
InChi Key: FFLPJEKRAYZAMU-UHFFFAOYSA-N
SMILES: Cl.NC(=N)C1=CC(Cl)=CC(Cl)=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results