accessibility menu, dialog, popup

UserName

SOOBr

CAS RN 2297-65-6

IUPAC Name: benzenesulfonyl bromide
Synonyms: benzenesulfonyl bromide
Molecular Weight (g/mol): 221.07
Molecular Formula: C6H5BrO2S
InChi Key: CGWWQPZGKPHLBU-UHFFFAOYSA-N
SMILES: BrS(=O)(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results