accessibility menu, dialog, popup

UserName

CAS RN 22927-13-5

H3CO

CAS RN 22927-13-5

IUPAC Name: 2-ethylbenzaldehyde
Synonyms: 2-ethylbenzaldehyde
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: NTWBHJYRDKBGBR-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results