CAS RN 22916-47-8
Miconazole, 97%, Thermo Scientific™
CAS: 22916-47-8 Molecular Formula: C18H14Cl4N2O Molecular Weight (g/mol): 416.12 MDL Number: MFCD00216019 InChI Key: BYBLEWFAAKGYCD-UHFFFAOYNA-N Synonym: miconazole,monistat,monistat iv,daktarin iv,miconazolo,miconazolum,miconazol,minostate,dactarin,monistat-derm PubChem CID: 4189 ChEBI: CHEBI:82892 IUPAC Name: 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole SMILES: ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1
Miconazole Free Base MP Biomedicals
CAS: 22916-47-8 Molecular Formula: C18H14Cl4N2O Molecular Weight (g/mol): 416.12 MDL Number: MFCD00216019 InChI Key: BYBLEWFAAKGYCD-UHFFFAOYNA-N Synonym: miconazole,monistat,monistat iv,daktarin iv,miconazolo,miconazolum,miconazol,minostate,dactarin,monistat-derm PubChem CID: 4189 ChEBI: CHEBI:82892 IUPAC Name: 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole SMILES: ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1
Miconazole, TRC
CAS: 22916-47-8 Molecular Formula: C18 H14 Cl4 N2 O Molecular Weight (g/mol): 416.13 Synonym: Imidazole, 1-[2,4-dichloro-β-[(2,4-dichlorobenzyl)oxy]phenethyl]- (8CI),1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole,(±)-Miconazole,Daktanol,Daktarin IV,Florid-F,Lauriad,Loramyc,MJR 1762,Miconazole,Monistat IV,NSC 170986,R 18134,Zimybase IUPAC Name: 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole SMILES: Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)c(Cl)c1
Miconazole, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Miconazole, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Miconazole (Free Base), Fisher BioReagents
CAS: 22916-47-8 Molecular Formula: C18H14Cl4N2O Molecular Weight (g/mol): 416.12 MDL Number: MFCD00216019 InChI Key: BYBLEWFAAKGYCD-UHFFFAOYNA-N Synonym: 1-[2, 4-Dichloro-β-([2, 4-dichloro-benzyl]oxy)phenethyl]imidazole PubChem CID: 4189 ChEBI: CHEBI:82892 SMILES: ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1