CAS RN 229018-17-1
CAS RN 229018-17-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxymelatonin, TRC
CAS: 229018-17-1 Molecular Formula: C13H16N2O3 Molecular Weight (g/mol): 248.28 Synonym: N-[2-(2-Hydroxy-5-methoxy-1H-indol-3-yl)ethyl]acetamide IUPAC Name: N-[2-(2-hydroxy-5-methoxy-1H-indol-3-yl)ethyl]acetamide SMILES: CC(NCCC1=C(O)NC2=C1C=C(OC)C=C2)=O