CAS RN 22884-29-3
IUPAC Name:
(2-methylpropyl)triphenylphosphanium bromide
Synonyms:
(2-methylpropyl)triphenylphosphanium bromide
Molecular Weight (g/mol):
399.31
Molecular Formula:
C22H24BrP
InChi Key:
VIKPDPHNYHUZQW-UHFFFAOYSA-M
SMILES:
[Br-].CC(C)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Isobutyltriphenylphosphonium bromide, 98+%, Thermo Scientific™
CAS: 22884-29-3 Molecular Formula: C22H24BrP Molecular Weight (g/mol): 399.31 MDL Number: MFCD00031593 InChI Key: VIKPDPHNYHUZQW-UHFFFAOYSA-M Synonym: isobutyltriphenylphosphonium bromide,2-methylpropyl triphenyl phosphanium bromide,phosphonium, 2-methylpropyl triphenyl-, bromide,2-methylpropyl triphenylphosphanium bromide,acmc-1citv,isobutyltriphenylphosphoniumbromide,isobutyl triphenylphosphonium bromide,2-methylpropyl triphenyl phosphonium bromide,isobutyl triphenyl phosphonium bromide,phosphonium, 2-methylpropyl triphenyl-,bromide 1:1 PubChem CID: 2724571 SMILES: [Br-].CC(C)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1