CAS RN 22867-74-9
CAS RN 22867-74-9
IUPAC Name:
7-ethyl-1H-indole
Synonyms:
7-ethyl-1H-indole
Molecular Weight (g/mol):
145.21
Molecular Formula:
C10H11N
InChi Key:
PIIZLMYXLGYWTN-UHFFFAOYSA-N
SMILES:
CCC1=C2NC=CC2=CC=C1
1
–
2
of
2
results
1
7-Ethylindole, 98+%
CAS: 22867-74-9 Molecular Formula: C10H11N Molecular Weight (g/mol): 145.21 MDL Number: MFCD00143514 InChI Key: PIIZLMYXLGYWTN-UHFFFAOYSA-N Synonym: 7-ethylindole,1h-indole, 7-ethyl,7-ethyl indole,pubchem7338,acmc-1cfby,7-ethylindole gc PubChem CID: 2724711 IUPAC Name: 7-ethyl-1H-indole SMILES: CCC1=C2NC=CC2=CC=C1
2
7-Ethylindole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.