CAS RN 228418-45-9
CAS RN 228418-45-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Methyl 2-Nitro-4-(trifluoromethyl)benzoate, TRC
CAS: 228418-45-9 Molecular Formula: C9H6F3NO4 Molecular Weight (g/mol): 249.15 Synonym: 2-Nitro-4-trifluoromethylbenzoic Acid Methyl Ester,Methyl 2-Nitro-4-trifluoromethylbenzoate IUPAC Name: methyl 2-nitro-4-(trifluoromethyl)benzoate SMILES: COC(=O)c1ccc(cc1[N+](=O)[O-])C(F)(F)F